Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Bromomethyl-3-nitrobenzoic acid, 97%

Catalog No. AAB2525503
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2525503 1 g
AAB2525506 5 g
AAB2525514 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2525503 Supplier Thermo Scientific Chemicals Supplier No. B2525503
Only null left
Add to Cart

CAS: 55715-03-2 | C8H6BrNO4 | 260.043 g/mol

4-Bromomethyl-3-nitrobenzoic acid is used as a reactant in the synthesis of 4-bromomethyl-3-nitrobenzoic acid succinimide ester, 4-((2-(hydroxymethyl)phenyl amino)methyl)-3-nitrobenzoic acid, 4-(2-hydroxyethylmercaptylmethyl)-3-nitrobenzoic acid. It is also used as a thiol photo-deprotection reagent, a starting material in the synthesis of 2H-indazole based library using parallel solution-phase methods.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Bromomethyl-3-nitrobenzoic acid is used as a reactant in the synthesis of 4-bromomethyl-3-nitrobenzoic acid succinimide ester, 4-((2-(hydroxymethyl)phenyl amino)methyl)-3-nitrobenzoic acid, 4-(2-hydroxyethylmercaptylmethyl)-3-nitrobenzoic acid. It is also used as a thiol photo-deprotection reagent, a starting material in the synthesis of 2H-indazole based library using parallel solution-phase methods.

Solubility
Soluble in DMF and dichloromethane. Insoluble in water.

Notes
Store in a cool, dry conditions, in well sealed conditions. Store away from oxidizing agents, bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 55715-03-2
Molecular Formula C8H6BrNO4
Molecular Weight (g/mol) 260.043
MDL Number MFCD00216575
InChI Key QMAHVAFURJBOFV-UHFFFAOYSA-N
Synonym 4-bromomethyl-3-nitrobenzoic acid, 4-bromomethyl-3-nitrobenzoicacid, benzoic acid, 4-bromomethyl-3-nitro, 3-nitro-4-bromomethylbenzoic acid, bnba, acmc-1auor, ksc495i9f, 3-nitro-4-bromomethyl benzoic acid, 4-bromomethyl-3-nitro benzoic acid
PubChem CID 132386
SMILES C1=CC(=C(C=C1C(=O)O)[N+](=O)[O-])CBr

Specifications

Melting Point 128°C to 133°C
Quantity 1 g
UN Number UN3261
Solubility Information Soluble in DMF and dichloromethane. Insoluble in water.
Formula Weight 260.05
Percent Purity 97%
Chemical Name or Material 4-Bromomethyl-3-nitrobenzoic acid
Hazard Category H314
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.