Learn More
4-Bromomethyl-3-nitrobenzoic acid, 97%
Description
4-Bromomethyl-3-nitrobenzoic acid is used as a reactant in the synthesis of 4-bromomethyl-3-nitrobenzoic acid succinimide ester, 4-((2-(hydroxymethyl)phenyl amino)methyl)-3-nitrobenzoic acid, 4-(2-hydroxyethylmercaptylmethyl)-3-nitrobenzoic acid. It is also used as a thiol photo-deprotection reagent, a starting material in the synthesis of 2H-indazole based library using parallel solution-phase methods.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
4-Bromomethyl-3-nitrobenzoic acid is used as a reactant in the synthesis of 4-bromomethyl-3-nitrobenzoic acid succinimide ester, 4-((2-(hydroxymethyl)phenyl amino)methyl)-3-nitrobenzoic acid, 4-(2-hydroxyethylmercaptylmethyl)-3-nitrobenzoic acid. It is also used as a thiol photo-deprotection reagent, a starting material in the synthesis of 2H-indazole based library using parallel solution-phase methods.
Solubility
Soluble in DMF and dichloromethane. Insoluble in water.
Notes
Store in a cool, dry conditions, in well sealed conditions. Store away from oxidizing agents, bases.
Specifications
Specifications
| Melting Point | 128°C to 133°C |
| Quantity | 5 g |
| UN Number | UN3261 |
| Solubility Information | Soluble in DMF and dichloromethane. Insoluble in water. |
| Formula Weight | 260.05 |
| Percent Purity | 97% |
| Chemical Name or Material | 4-Bromomethyl-3-nitrobenzoic acid |
Safety and Handling
| Hazard Category | H314 |
| Hazard Statement | GHS H Statement H314-H318 Causes severe skin burns and eye damage. Causes serious eye damage. |
| Precautionary Statement | P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c |
| DOTInformation | Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S. |
| TSCA | No |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.