Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Bromomethyl-3-nitrobenzoic acid, 97%, Thermo Scientific™

Catalog No. AC334060010
Change view
Click to view available options
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
AC334060010 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC334060010 Supplier Thermo Scientific Chemicals Supplier No. 334060010

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 55715-03-2
CAS Min % 96.0
CAS Max % 100.0
Infrared Spectrum Authentic
Assay Percent Range 97%
Molecular Formula C8H6BrNO4
Quantity 1g
MDL Number MFCD00216575
Synonym 4-bromomethyl-3-nitrobenzoic acid, 4-bromomethyl-3-nitrobenzoicacid, benzoic acid, 4-bromomethyl-3-nitro, 3-nitro-4-bromomethylbenzoic acid, bnba, acmc-1auor, ksc495i9f, 3-nitro-4-bromomethyl benzoic acid, 4-bromomethyl-3-nitro benzoic acid
InChI Key QMAHVAFURJBOFV-UHFFFAOYSA-N
SMILES C1=CC(=C(C=C1C(=O)O)[N+](=O)[O-])CBr
Molecular Weight (g/mol) 260.043
PubChem CID 132386
Formula Weight 260.04
Percent Purity ≥96.0%
Chemical Name or Material 4-Bromomethyl-3-nitrobenzoic acid
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.