Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Bromomethyl benzoyl bromide, 96%, Thermo Scientific™

Catalog No. AAH3768214
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3768214 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH3768214 Supplier Thermo Scientific Chemicals Supplier No. H3768214

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 876-07-3
Molecular Formula C8H6Br2O
Molecular Weight (g/mol) 277.94
MDL Number MFCD00045026
InChI Key VUYGJYAPDGKPBF-UHFFFAOYSA-N
Synonym 4-bromomethyl benzoyl bromide, acmc-20ammq, 4-bromomethyl benzoylbromide, 4-bromomethyl-benzoyl bromide, benzoyl bromide, 4-bromomethyl, p-bromomethyl benzoyl bromide, 4-bromomethyl benzoyl bromide 96
PubChem CID 4640671
IUPAC Name 4-(bromomethyl)benzoyl bromide
SMILES BrCC1=CC=C(C=C1)C(Br)=O

Specifications

Melting Point 57°C to 61°C
Color White
Boiling Point 100°C to 112°C (0.5 mmHg)
Linear Formula BrCH2C6H4C(O)Br
Quantity 25 g
UN Number UN3261
Sensitivity Air Sensitive
Formula Weight 277.94
Percent Purity 96%
Physical Form Fused Solid
Chemical Name or Material 4-Bromomethyl benzoyl bromide
Show More Show Less
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packaging Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures; Store under Argon
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.