Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-(Bromomethyl)tetrahydropyran, 97%, Thermo Scientific™

Catalog No. CC29908EA
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Packaging:
Amber Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC29908EA 10 g Amber Glass Bottle
CC29908DA 1 g Amber Glass Bottle
CC29908DE 5 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. CC29908EA Supplier Thermo Scientific Chemicals Supplier No. CC29908EA
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 125552-89-8
Molecular Formula C6H11BrO
Molecular Weight (g/mol) 179.057
InChI Key LMOOYAKLEOGKJR-UHFFFAOYSA-N
Synonym 4-bromomethyl tetrahydropyran, 4-bromomethyltetrahydropyran, 4-bromomethyl oxane, 4-bromomethyl tetrahydro-2h-pyran, 4-bromomethyl-tetrahydro-pyran, 4-bromomethyl tetrahydro-pyran, 4-bromomethyl-tetrahydro-2h-pyran, 4-bromomethyl-tetrahydropyran, 2h-pyran, 4-bromomethyl tetrahydro
PubChem CID 2773286
IUPAC Name 4-(bromomethyl)oxane
SMILES C1COCCC1CBr

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 10 g
Formula Weight 179.06
Percent Purity 97%
Chemical Name or Material 4-(Bromomethyl)tetrahydropyran
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.