Learn More
AdipoGen 4-Bromomethylphenyl ITC
SDP
Catalog No. 502561684
Encompass_Preferred
Chemical. CAS 155863-32-4. Formula C8H6BrNS. MW 228.1. Synthetic. Hetero-bifunctional linker specially for the quaternisation of pyridyl-nitrogen in order to introduce an amine or thiol reactive group. Synthetic building block for the preparation of fluorescent labels.
Catalog No. 50-256-1684 Supplier AdipoGen Supplier No. CDXB0091M025
Chemical. CAS 155863-32-4. Formula C8H6BrNS. MW 228.1. Synthetic. Hetero-bifunctional linker specially for the quaternisation of pyridyl-nitrogen in order to introduce an amine or thiol reactive group. Synthetic building block for the preparation of fluorescent labels.