Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 4-Bromophenethyl alcohol

Catalog No. 18343110G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
18-343-110G 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18-343-110G Supplier Sigma Aldrich Supplier No. 18343110G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

4-Bromophenethyl alcohol

Specifications

CAS 4654-39-1
Density 1.436 g/mL (at 25°C (literature))
Boiling Point 138°C (9 mmHg)
Molecular Formula C8H9BrO
Refractive Index n20/D 1.573 (literature)
Linear Formula BrC6H4CH2CH2OH
Quantity 250 mg
MDL Number MFCD00002897
Synonym 2-(4-Bromophenyl)ethanol
Molecular Weight (g/mol) 201.06
Percent Purity 99%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.