Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Bromophenyl Cyclopropyl Ketone 95.0+%, TCI America™
SDP

Catalog No. B40761G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B40761G 1 g
B40765G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B40761G Supplier TCI America Supplier No. B40761G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 6952-89-2
Molecular Formula C10H9BrO
Molecular Weight (g/mol) 225.085
MDL Number MFCD00019227
InChI Key QTHHOINSCNBYQO-UHFFFAOYSA-N
Synonym 4-bromophenyl cyclopropyl methanone, 4-bromophenyl cyclopropyl ketone, 4-bromophenyl cyclopropylmethanone, methanone, 4-bromophenyl cyclopropyl, 4-bromo-cyclopropylcarbonylbenzene, acmc-1bag8, 4-bromobenzoyl cyclopropane, bromophenylcyclopropylmethanone
PubChem CID 81394
IUPAC Name (4-bromophenyl)-cyclopropylmethanone
SMILES C1CC1C(=O)C2=CC=C(C=C2)Br

Specifications

Melting Point 43°C
Color White-Yellow
Boiling Point 204°C
Quantity 1 g
Formula Weight 225.09
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 4-Bromophenyl Cyclopropyl Ketone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.