Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(4-Bromophenyl)diphenylphosphine Oxide 98.0+%, TCI America™
SDP

Catalog No. B54591G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B54591G 1 g
B54595G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B54591G Supplier TCI America Supplier No. B54591G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 5525-40-6
Molecular Formula C18H14BrOP
Molecular Weight (g/mol) 357.187
MDL Number MFCD00411450
InChI Key PCYBTUUJXASDIX-UHFFFAOYSA-N
PubChem CID 11268306
IUPAC Name 1-bromo-4-diphenylphosphorylbenzene
SMILES C1=CC=C(C=C1)P(=O)(C2=CC=CC=C2)C3=CC=C(C=C3)Br

Specifications

Melting Point 156°C
Color White-Yellow
Quantity 1 g
Formula Weight 357.19
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material (4-Bromophenyl)diphenylphosphine Oxide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.