Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Bromophenylacetic acid, 99%

Catalog No. p-7022506
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1532514 25 g
AAA1532522 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1532514 Supplier Thermo Scientific Chemicals Supplier No. A1532514
Only null left
Add to Cart

CAS: 1878-68-8 | C8H7BrO2 | 215.05 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1878-68-8
Molecular Formula C8H7BrO2
Molecular Weight (g/mol) 215.05
MDL Number MFCD00004342
InChI Key QOWSWEBLNVACCL-UHFFFAOYSA-N
Synonym 4-bromophenylacetic acid, 2-4-bromophenyl acetic acid, 4-bromophenyl acetic acid, p-bromophenylacetic acid, 4-bromobenzeneacetic acid, benzeneacetic acid, 4-bromo, p-bromophenyl acetic acid, acetic acid, p-bromophenyl, p-bromphenylessigsaure german
PubChem CID 74654
ChEBI CHEBI:1790
IUPAC Name 2-(4-bromophenyl)acetic acid
SMILES OC(=O)CC1=CC=C(Br)C=C1

Specifications

Melting Point 115°C to 119°C
Quantity 25 g
Beilstein 972146
Formula Weight 215.05
Percent Purity 99%
Chemical Name or Material 4-Bromophenylacetic acid
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 217-523-7
RTECSNumber AF6325000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.