Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 4-Bromopyrazole | 2075-45-8 | 99.79% | C₃H₃BrN₂ | 100 G
SDP

Catalog No. 5000434561
Encompass_Preferred

4-Bromopyrazole is a biochemical reagent that can be used as a biological material or organic compound for life science related research. It is for research use only.

  • High purity: 99.79%
  • Appearance: Off-white to light yellow solid
  • Molecular weight: 146.97
  • For research use only

Catalog No. 50-004-34561 Supplier Medchemexpress LLC Supplier No. HY78243100G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

4-Bromopyrazole is a biochemical reagent that can be used as a biological material or organic compound for life science related research. It is for research use only.

  • High purity: 99.79%
  • Appearance: Off-white to light yellow solid
  • Molecular weight: 146.97
  • For research use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.