Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Bromopyridine Hydrobromide 98.0+%, TCI America™
SDP

Catalog No. B11715G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B11715G 5 g
B117125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B11715G Supplier TCI America Supplier No. B11715G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 74129-11-6
Molecular Formula C5H5Br2N
Molecular Weight (g/mol) 238.91
MDL Number MFCD00060195
InChI Key SMSVBXDZZQOTGB-UHFFFAOYSA-N
PubChem CID 44630127
IUPAC Name 4-bromopyridine hydrobromide
SMILES Br.BrC1=CC=NC=C1

Specifications

Color White-Yellow
Quantity 5 g
UN Number 2811
Formula Weight 238.91
Percent Purity ≥98.0% (T)
Grade Laboratory
Physical Form Crystalline Powder
Chemical Name or Material 4-Bromopyridine Hydrobromide
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.