Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 4-Bromoresorcinol

Catalog No. B8060725G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B8060725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B8060725G Supplier Sigma Aldrich Supplier No. B8060725G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Reactant for preparation of:
  • Inhibitors of nod1-induced nuclear factor-αb activation
  • Sphingosine-1-phosphate 4(s1p4) receptor antagonists
  • Cytotoxic cell cycle inhibitors
  • 2-aminopyridines
  • Pet agent for imaging of protein kinase c (pkc)
  • Sodium-dependent glucose co-transporter 2 (sglt2) inhibitors for the management of hyperglycemia in diabetes
  • α4α2-nicotinic acetylcholine receptor-selective partial agonists
  • Mglu4 positive allosteric modulators
  • Bacterial biofilm inhibitors
  • Serotonin 5-ht6 receptor antagonistsreagentplus is a registered trademark of sigma-aldrich co. llc

Specifications

CAS 6626-15-9
Melting Point 97°C to 100°C (lit.)
Boiling Point 150°C (12 mmHg)
Molecular Formula C6H5BrO2
Linear Formula BrC6H3-1,3-(OH)2
MDL Number MFCD00002272
Quantity 25 g
Molecular Weight (g/mol) 189.01
Percent Purity 97%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.