Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4-Bromoresorcinol, 98%

Click to view available options
Quantity:
5g
25g
6626-15-9
C6H5BrO2
189.008
MFCD00002272
MPCCNXGZCOXPMG-UHFFFAOYSA-N
4-bromoresorcinol,1-bromo-2,4-dihydroxybenzene,1,3-benzenediol, 4-bromo,4-bromo-1,3-dihydroxybenzene,4-bromo-benzene-1,3-diol,4-bromo-resorcinol,resorcinol, 4-bromo,acmc-209nuf,2,4-dihydroxybromobenzene,4-bromoresorcinol
81105
4-bromobenzene-1,3-diol
C1=CC(=C(C=C1O)O)Br
6626-15-9
C6H5BrO2
189.008
MFCD00002272
MPCCNXGZCOXPMG-UHFFFAOYSA-N
4-bromoresorcinol,1-bromo-2,4-dihydroxybenzene,1,3-benzenediol, 4-bromo,4-bromo-1,3-dihydroxybenzene,4-bromo-benzene-1,3-diol,4-bromo-resorcinol,resorcinol, 4-bromo,acmc-209nuf,2,4-dihydroxybromobenzene,4-bromoresorcinol
81105
4-bromobenzene-1,3-diol
C1=CC(=C(C=C1O)O)Br
Applications
It is commonly used as an organic reagent and pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
Color Pink
Odor Odorless
Melting Point 97°C to 100°C
Boiling Point 150°C (12mmHg)
Quantity 5g
Beilstein 1861280
Formula Weight 189.01
Physical Form Powder
Percent Purity 98%
Chemical Name or Material 4-Bromoresorcinol

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning