Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4'-Bromosalicylanilide 98.0+%, TCI America™
SDP

Catalog No. B09195G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B09195G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B09195G Supplier TCI America Supplier No. B09195G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 2627-77-2
Molecular Formula C13H10BrNO2
Molecular Weight (g/mol) 292.13
MDL Number MFCD00059615
InChI Key DKQMTUHJJPFJRL-UHFFFAOYSA-N
Synonym 4′C-Bromo-2-hydroxybenzanilide
PubChem CID 75817
IUPAC Name N-(4-bromophenyl)-2-hydroxybenzamide
SMILES OC1=CC=CC=C1C(=O)NC1=CC=C(Br)C=C1

Specifications

Melting Point 172°C
Quantity 5 g
Formula Weight 292.13
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 4′-Bromosalicylanilide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.