Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 4-Bromostyrene

Catalog No. 12414110G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
12-414-110G 10 g
12-414-125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 12-414-110G Supplier Sigma Aldrich Supplier No. 12414110G
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • 4-Bromostyrene is a para-halogenated styrene derivative. The proton spectra of 4-bromostyrene exhibits dipolar couplings consistent with planar ground state structures, only if the torsional motion of lowest frequency occurs at about 80cm-1. It undergoes Heck reaction with 2-bromo-6-methoxynaphthalene in the presence of sodium acetate and Hermann's catalyst in N, N-dimethylacetamide to afford diarylethene. 4-Bromostyrene was used in the following studies:
  • Structure-activity relationships (SAR) study of the chemical and biochemical properties of the vinyl group of styrene.
  • Synthesis of silsesquioxanes (SQ) having 4-bromostyrenyl substituents.
  • To investigate the photochemical growth of Br-terminated self-assembled monolayers (SAMs) on Si(111).
  • Synthesis of poly(1,4-phenylenevinylene) via Heck reaction.
  • Synthesis of nitroolefins via alkene cross-metathesis.

Specifications

CAS 2039-82-9
Density 1.4 g/mL (at 25°C)
Boiling Point 89°C (6 mmHg)
Molecular Formula C8H7Br
Refractive Index n20/D 1.594 (literature)
Linear Formula H2C=CHC6H4Br
MDL Number MFCD00000110
Quantity 10 g
Synonym 1-(4-Bromophenyl)ethylene; 1-Bromo-4-ethenylbenzene; 1-Bromo-4-vinylbenzene; 4-Vinyl-1-bromobenzene; p -Bromostyrene
Molecular Weight (g/mol) 183.05
Percent Purity 97%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.