Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Bromothiophene-3-carboxaldehyde 97.0+%, TCI America™
SDP

Catalog No. B41261G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B41261G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B41261G Supplier TCI America Supplier No. B41261G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 18791-78-1
Molecular Formula C5H3BrOS
Molecular Weight (g/mol) 191.04
MDL Number MFCD01318960
InChI Key ZDVHJQOCVBTMOF-UHFFFAOYSA-N
Synonym 4-Bromo-3-formylthiophene
PubChem CID 2764207
IUPAC Name 4-bromothiophene-3-carbaldehyde
SMILES BrC1=CSC=C1C=O

Specifications

Color Yellow
Boiling Point 120°C
Quantity 1 g
Formula Weight 191.04
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-Bromothiophene-3-carboxaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.