Learn More
Bellco Glass, Inc. 4-Bottom Baffles Flask,500mL 38mm Delong Neck
SDP
Catalog No. 501945547
Encompass_Preferred
Manufactured from Type 1 Class A Borosilicate Glass. Shake flasks are the basic tool for growth studies of aerobic microbial cultures. They have rigidly controlled baffle angles and depths to promote optimal oxygen transfer while reducing vortex formation. Designed for all orbital shakers.. MADE IN THE USASpecifications:Type 1 Class A Borosilicate GlassSize: 500mlDimensions: 100x203mmClosure Size: 38Case Quantity: 12
Catalog No. 50-194-5547 Supplier Bellco Glass, Inc. Supplier No. 254700500
Manufactured from Type 1 Class A Borosilicate Glass. Shake flasks are the basic tool for growth studies of aerobic microbial cultures. They have rigidly controlled baffle angles and depths to promote optimal oxygen transfer while reducing vortex formation. Designed for all orbital shakers.. MADE IN THE USASpecifications:Type 1 Class A Borosilicate GlassSize: 500mlDimensions: 100x203mmClosure Size: 38Case Quantity: 12