Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4'-Butoxybenzylidene-4-cyanoaniline 98.0+%, TCI America™
SDP

Catalog No. B02521G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B02521G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B02521G Supplier TCI America Supplier No. B02521G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 36405-17-1
Molecular Formula C18H18N2O
Molecular Weight (g/mol) 278.355
MDL Number MFCD00063742
InChI Key JWLPZJPDDBBWQD-UHFFFAOYSA-N
Synonym 4-(4-Butoxybenzylideneamino)benzonitrile
PubChem CID 118967
IUPAC Name 4-[(4-butoxyphenyl)methylideneamino]benzonitrile
SMILES CCCCOC1=CC=C(C=C1)C=NC2=CC=C(C=C2)C#N

Specifications

Color Yellow
Quantity 1 g
Formula Weight 278.36
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 4′-Butoxybenzylidene-4-cyanoaniline
EINECSNumber (3)-1840
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.