Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 4-Butoxyphenol | 122-94-1 | 166.22 | 50 G
SDP

Catalog No. 5000402054
Encompass_Preferred

4-Butoxyphenol is a biochemical reagent that can be used as a biological material or organic compound for life science-related research. It is an organic compound.

  • For research use only.
  • Appearance: Solid.
  • Color: Light brown to brown.
  • Shipping: Room temperature in continental US; may vary elsewhere.
  • Storage: 4°C, stored under nitrogen. In solvent: -80°C for 6 months; -20°C for 1 month (stored under nitrogen).

Catalog No. 50-004-02054 Supplier Medchemexpress LLC Supplier No. HY3027150G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

4-Butoxyphenol is a biochemical reagent that can be used as a biological material or organic compound for life science-related research. It is an organic compound.

  • For research use only.
  • Appearance: Solid.
  • Color: Light brown to brown.
  • Shipping: Room temperature in continental US; may vary elsewhere.
  • Storage: 4°C, stored under nitrogen. In solvent: -80°C for 6 months; -20°C for 1 month (stored under nitrogen).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.