Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Butoxyphenylboronic Acid (contains varying amounts of Anhydride), TCI America™
SDP

Catalog No. B47275G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B4727-5G 5 g
B47271G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B4727-5G Supplier TCI America Supplier No. B47275G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 105365-51-3
Molecular Formula C10H15BO3
MDL Number MFCD03427054
InChI Key QUPFQMXWFNJUNJ-UHFFFAOYSA-N
PubChem CID 3836310
IUPAC Name (4-butoxyphenyl)boronic acid

Specifications

Melting Point 108°C
Color White
Quantity 5 g
Synonym 4-n-butoxyphenylboronic acid, 4-butoxyphenyl boronic acid, 4-n-butoxyphenyl boronic acid, p-butoxyphenylboronic acid, 4-butyloxyphenylboronic acid
SMILES B(C1=CC=C(C=C1)OCCCC)(O)O
Molecular Weight (g/mol) 194.037
Formula Weight 194.04
Physical Form Crystalline Powder
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.