Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Butyl-2-methylaniline, Tech., Thermo Scientific™

Catalog No. SB01607DA
Change view
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
SB01607DA 1 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. SB01607DA Supplier Thermo Scientific Chemicals Supplier No. SB01607DA
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 72072-16-3
Molecular Formula C11H17N
Molecular Weight (g/mol) 163.264
MDL Number MFCD00190660
InChI Key JTXOXRXZCAMPHL-UHFFFAOYSA-N
PubChem CID 577772
IUPAC Name 4-butyl-2-methylaniline
SMILES CCCCC1=CC(=C(C=C1)N)C

Specifications

Packaging Amber Glass Bottle
Quantity 1 g
Synonym benzenamine,4-butyl-2-methyl, 2-methyl-4-butylaniline, 4-butyl-2-methyl-aniline, 4-n-butyl-2-methylaniline, 4-butyl-2-methylphenylamine, #, benzenamine, 4-butyl-2-methyl
Formula Weight 163.26
Percent Purity Tech.
Grade Technical
Chemical Name or Material 4-Butyl-2-methylaniline
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.