Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-(Butylamino)-1-butanol 99.0+%, TCI America™
SDP

Catalog No. B095425ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B095425ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B095425ML Supplier TCI America Supplier No. B095425ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 4543-95-7
Molecular Formula C8H19NO
Molecular Weight (g/mol) 145.246
MDL Number MFCD00059007
InChI Key OPUJAKVDYGQVHP-UHFFFAOYSA-N
Synonym N-Butyl-4-hydroxybutylamine
PubChem CID 20669
IUPAC Name 4-(butylamino)butan-1-ol
SMILES CCCCNCCCCO

Specifications

Color Yellow
Boiling Point 132°C
Quantity 25 mL
Formula Weight 145.25
Percent Purity ≥99.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 4-(Butylamino)-1-butanol
RTECSNumber EL0900000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.