Learn More
Medchemexpress LLC 4-(Butylamino)benzoic acid-d7 | 1219803-44-7 | 99.7% | 200.29 | 1 MG
SDP
Catalog No. 5000404417
Encompass_Preferred
4-(Butylamino)benzoic acid-d7 is a deuterium labeled variant of 4-(Butylamino)benzoic acid. It is suitable for research applications and not intended for patient use. This compound is valuable as a tracer and internal standard for quantitative analysis.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-004-04417 Supplier Medchemexpress LLC Supplier No. HY41463S1MG
4-(Butylamino)benzoic acid-d7 is a deuterium labeled variant of 4-(Butylamino)benzoic acid. It is suitable for research applications and not intended for patient use. This compound is valuable as a tracer and internal standard for quantitative analysis.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS