Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 4-Butylaniline

Catalog No. 112666500G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-266-6500G 500 g
11-266-6100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11-266-6500G Supplier Sigma Aldrich Supplier No. 112666500G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

4-Butylaniline is used in the fabrication of RP/ion-exchange, mixed-mode, monolithic materials for capillary LC.4-Butylaniline is a mammalian retinoid cycle inhibitor that reversibly suppresses the recovery of the outward R(2) component of ERC (early receptor current) from Vitamin A and 11-cis-retinal-loaded cells.

Specifications

CAS 104-13-2
Density 0.945 g/mL (at 25°C (literature))
Boiling Point 133°C to 134°C (14 mmHg)
Molecular Formula C10H15N
Linear Formula CH3(CH2)3C6H4NH2
Refractive Index n20/D 1.535 (literature)
MDL Number MFCD00007925
Quantity 500 g
Molecular Weight (g/mol) 149.23
Percent Purity 97%
Biological Activity Respiratory System
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.