Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Butylbenzaldehyde Diethyl Acetal 97.0+%, TCI America™
SDP

Catalog No. B12225ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B12225ML 5 mL
B122225ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B12225ML Supplier TCI America Supplier No. B12225ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 83803-80-9
Molecular Formula C15H24O2
Molecular Weight (g/mol) 236.36
MDL Number MFCD00009466
InChI Key VZIJDIWCWDYVFM-UHFFFAOYSA-N
Synonym 1-butyl-4-diethoxymethyl benzene, 4-butylbenzaldehyde diethyl acetal, 1-butyl-4-diethoxymethyl-benzene, benzene,1-butyl-4-diethoxymethyl, 4-butylphenyl diethoxymethane, 4-butylbenzaldehydediethylacetal, acmc-209ptb, 4-butyl-, a,, a-diethoxytoluene, 4-butylbenzaldehyde diethylacetal, 4-n-butylbenzaldehyde diethyl acetal
PubChem CID 605679
IUPAC Name 1-butyl-4-(diethoxymethyl)benzene
SMILES CCCCC1=CC=C(C=C1)C(OCC)OCC

Specifications

Color Yellow
Quantity 5 mL
Formula Weight 236.36
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-Butylbenzaldehyde Diethyl Acetal
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.