Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Butylbenzene-1-sulfonyl chloride, 97%, Thermo Scientific™

Catalog No. BTB10243EA
Change view
Click to view available options
Quantity:
10 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BTB10243EA 10 g
BTB10243EB 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BTB10243EA Supplier Thermo Scientific Chemicals Supplier No. BTB10243EA
Add to Cart

Chemical Identifiers

CAS 54997-92-1
Molecular Formula C10H13ClO2S
Molecular Weight (g/mol) 232.72
MDL Number MFCD00173908
InChI Key OVFZELSNOHIDEF-UHFFFAOYSA-N
Synonym 4-butylbenzene-1-sulfonyl chloride, 4-n-butylbenzenesulfonyl chloride, 4-n-butyl benzenesulphonyl chloride, 4-butyl-benzenesulfonyl chloride, 4-n-butylphenylsulfonyl chloride, 4-n-butylbenzenesulphonyl chloride, benzenesulfonyl chloride, 4-butyl, 4-butylphenyl chlorosulfone, pubchem5484, 4-n-butylbenzenesulfonylchloride
PubChem CID 2737352
SMILES CCCCC1=CC=C(C=C1)S(Cl)(=O)=O

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Linear Formula CH3(CH2)3C6H4SO2Cl
Quantity 10 g
IUPAC Name 4-butylbenzenesulfonyl chloride
Formula Weight 232.73
Percent Purity 97%
Chemical Name or Material 4-Butylbenzene-1-sulfonyl chloride
Hazard Category Danger
Hazard Statement GHS P Statement Causes severe skin burns and eye damage. Contact with water liberates toxic gas. Reacts violently with water.
Precautionary Statement GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF SWALLOWED: rinse mouth. Do NOT induce vomiting. Immediately call a POISON CENTER or doctor/physician. IF IN EYES: Rinse cau
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.