Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Butylbenzoic Acid 98.0+%, TCI America™
SDP

Catalog No. B02415G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B02415G 5 g
B024125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B02415G Supplier TCI America Supplier No. B02415G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 20651-71-2
Molecular Formula C11H14O2
Molecular Weight (g/mol) 178.231
MDL Number MFCD00002571
InChI Key JFKUBRAOUZEZSL-UHFFFAOYSA-N
Synonym 4-n-butylbenzoic acid, benzoic acid, 4-butyl, 4-butyl-benzoic acid, p-butylbenzoic acid, p-n-butylbenzoic acid, p-butyl benzoic acid, 4-butylbenzoicacid, p-n-butyl benzoic acid, acmc-209fcp, 4-n-butyl-benzoic acid
PubChem CID 88631
IUPAC Name 4-butylbenzoic acid
SMILES CCCCC1=CC=C(C=C1)C(=O)O

Specifications

Melting Point 100°C
Color White-Yellow
Quantity 5 g
Formula Weight 178.23
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Butylbenzoic Acid
EINECSNumber (3)-1338
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.