Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 4-Butylphenol-d5 | 1219795-04-6 | 98.3% | 155.25 g/mol | C10H9D5O | 1 MG
SDP

Catalog No. 5000283089
Encompass_Preferred

4-Butylphenol-d5 is the deuterium-labeled analogue of 4-butylphenol provided as a characterized reference material for analytical chemistry. It is intended for use as an internal standard, tracer, or reference compound in mass spectrometry, quantitative assays, and method development, supporting accurate quantitation and identification in complex matrices.

  • Deuterium-labeled internal standard for mass spectrometry and quantitative assays.
  • CAS number 1219795-04-6 for unambiguous identification.
  • Molecular weight 155.25 g/mol.
  • High purity (≈98.3%) suitable for analytical use; confirm with certificate of analysis.
  • Available in small packaged quantities (e.g., 1 mg) for analytical workflows.

Catalog No. 50-002-83089 Supplier Medchemexpress LLC Supplier No. HYW144683S1MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

4-Butylphenol-d5 is the deuterium-labeled analogue of 4-butylphenol provided as a characterized reference material for analytical chemistry. It is intended for use as an internal standard, tracer, or reference compound in mass spectrometry, quantitative assays, and method development, supporting accurate quantitation and identification in complex matrices.

  • Deuterium-labeled internal standard for mass spectrometry and quantitative assays.
  • CAS number 1219795-04-6 for unambiguous identification.
  • Molecular weight 155.25 g/mol.
  • High purity (≈98.3%) suitable for analytical use; confirm with certificate of analysis.
  • Available in small packaged quantities (e.g., 1 mg) for analytical workflows.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.