Learn More
Selleck Chemical LLC 4-Butylresorcinol S4997-25mg
SDP
Catalog No. 503112136
Encompass_Preferred
4-Butylresorcinol (4-n-Butylresorcinol Rucinol) is a potent inhibitor of tyrosinase and is used in cosmetics as a depigmenting agent
Catalog No. 50-311-2136 Supplier Selleck Chemical LLC Supplier No. S499725MG
4-Butylresorcinol (4-n-Butylresorcinol Rucinol) is a potent inhibitor of tyrosinase and is used in cosmetics as a depigmenting agent