Learn More
TARGETMOL CHEMICALS INC 4-Butylresorcinol 500MG
SDP
Catalog No. 502877243
Encompass_Preferred
Also available in 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes.4-Butylresorcinol (Butylresorcinol) is a potent inhibitor of tyrosinase(IC50 11.27 uM), and is used in cosmetics as a depigmenting agent. Purity 98%
Catalog No. 50-287-7243 Supplier TARGETMOL CHEMICALS INC Supplier No. T7125500MG
Also available in 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes.4-Butylresorcinol (Butylresorcinol) is a potent inhibitor of tyrosinase(IC50 11.27 uM), and is used in cosmetics as a depigmenting agent. Purity 98%