Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Butylresorcinol, ≥97.0% (GC), MilliporeSigma™ Supelco™

Catalog No. 111018579 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-8579 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-8579 Supplier MilliporeSigma Supelco Supplier No. 49797100MG
Add to Cart
Add to Cart

4-n-butylresorcinol is a derivative of resorcinol and a potent human tyrosinase inhibitor. It may be used to decrease skin irritation and is also known to inhibit melanin production.

Cleaning Products, Cosmetics, Food and Beverages and Personal Care
TRUSTED_SUSTAINABILITY

Specifications

CAS 18979-61-8
Flash Point 145°C
Linear Formula C10H14O2
MDL Number MFCD01684800
Synonym 4-Butyl-1,3-benzenediol
Type Analytical Standard
Quantity 100 mg
Formula Weight 166.22
Percent Purity ≥97.0% (GC)
Grade Analytical Standard
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.