Learn More
Sigma Aldrich 4-Butylthiophenylboronic acid
Shop All MilliporeSigma Sigma Organic Chemistry Products
Description
- Tert-butyl chloroacetate was used in the synthesis of:
- Imidazol-1-yl-acetic acid hydrochloride
- Cis-disubstituted aziridine ester via aza-darzens reaction
- 1,10-diaza-18-crown-6 based sensors bearing a coumarin fluorophore
Specifications
Specifications
| CAS | 151588-38-4 |
| Molecular Formula | C10H15BO2S |
| Linear Formula | C10H15BO2S |
| MDL Number | MFCD06801729 |
| Quantity | 100 g |
| Molecular Weight (g/mol) | 210.1 |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.