Learn More
Cayman Chemical 4-CABhydrochlorIde 10mg
SDP
Catalog No. 502954411
Encompass_Preferred
An analog of PCA; inhibits the reuptake of serotonin (IC50 = 330 nM) and dopamine (IC50 = 2.3 μM)
Catalog No. 50-295-4411 Supplier Cayman Chemical Supplier No. 1389310 MG
An analog of PCA; inhibits the reuptake of serotonin (IC50 = 330 nM) and dopamine (IC50 = 2.3 μM)