Learn More
Cayman Chemical 4-CAB (hydrochloride), Purity: ≥95%, CAS: 23292-08-2, 50 mg
SDP
Catalog No. 502954541
Encompass_Preferred
An analog of PCA; inhibits the reuptake of serotonin (IC50 = 330 nM) and dopamine (IC50 = 2.3 μM)
Catalog No. 50-295-4541 Supplier Cayman Chemical Supplier No. 1389350 MG
An analog of PCA; inhibits the reuptake of serotonin (IC50 = 330 nM) and dopamine (IC50 = 2.3 μM)