Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Carbamoyl-1-hexadecylpyridinium Chloride 96.0+%, TCI America™
SDP

Catalog No. C16315G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C16315G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C16315G Supplier TCI America Supplier No. C16315G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 377085-58-0
Molecular Formula C22H39ClN2O
Molecular Weight (g/mol) 383.017
MDL Number MFCD02093456
InChI Key AGQLSZMFKAEZAK-UHFFFAOYSA-N
PubChem CID 44630216
IUPAC Name 1-hexadecylpyridin-1-ium-4-carboxamide;chloride
SMILES CCCCCCCCCCCCCCCC[N+]1=CC=C(C=C1)C(=O)N.[Cl-]

Specifications

Quantity 5 g
Formula Weight 383.02
Percent Purity ≥96.0% (T)
Chemical Name or Material 4-Carbamoyl-1-hexadecylpyridinium Chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.