Learn More
World Precision Instrument #4 Carbide Ball Mill
SDP
Catalog No. 502521390
Encompass
This line-powered micro drill will make easy work of grinding, finishing, cutting and drilling bone, teeth and other material. The high-torque 35,000 rpm (maximum) motor is quiet and has minimal vibration which reduces wear on the motor and provides greater comfort for you. It also features a forward and reverse switch, “E Type” handpiece and handpiece holder. The handpiece has a removable nose cone that can be cleaned and sterilized. It accepts 3/32” and 2.33mm bur shanks. Unlike battery-powered drills, this unit maintains consistent power for the duration of use. The wide range of speeds allows you to control the amount of heat generation.
Catalog No. 50-252-1390 Supplier World Precision Instrument Supplier No. 501856
This line-powered micro drill will make easy work of grinding, finishing, cutting and drilling bone, teeth and other material. The high-torque 35,000 rpm (maximum) motor is quiet and has minimal vibration which reduces wear on the motor and provides greater comfort for you. It also features a forward and reverse switch, “E Type” handpiece and handpiece holder. The handpiece has a removable nose cone that can be cleaned and sterilized. It accepts 3/32” and 2.33mm bur shanks. Unlike battery-powered drills, this unit maintains consistent power for the duration of use. The wide range of speeds allows you to control the amount of heat generation.