Learn More
eMolecules​ 138-41-0 | 4-Carboxybenzenesulfonamide | Combi-Blocks | MFCD00007938 | 201.200 | C7H7NO4S | 97.000 | NS(=O)(=O)c1ccc(cc1)C(O)=O | 5g | 290689617
Catalog No. 501596784 Shop All eMolecules Inc. Products
Encompass_Preferred
4-Carboxybenzenesulfonamide | Combi-Blocks | 138-41-0 | MFCD00007938 | 201.200 | C7H7NO4S | 97.000 | NS(=O)(=O)c1ccc(cc1)C(O)=O | 5g | 290689617
Catalog No. 50-159-6784 Supplier eMolecules​ Supplier No. 290689617
4-Carboxybenzenesulfonamide | Combi-Blocks | 138-41-0 | MFCD00007938 | 201.200 | C7H7NO4S | 97.000 | NS(=O)(=O)c1ccc(cc1)C(O)=O | 5g | 290689617