Learn More
Cayman Chemical (4-Carboxybutyl-d4)triphenylphosphonium (bromide), Purity: ≥99% deuterated forms (d1-d4), CAS: 42932-63-8, 10 mg
SDP
Catalog No. 502978935
Encompass_Preferred
An internal standard for the quantification of (4-carboxybutyl)triphenylphosphonium by GC- or LC-MS
Catalog No. 50-297-8935 Supplier Cayman Chemical Supplier No. 3125410 MG
An internal standard for the quantification of (4-carboxybutyl)triphenylphosphonium by GC- or LC-MS