Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(4-Carboxybutyl)triphenylphosphonium Bromide 98%, Thermo Scientific™

Catalog No. AC169580500
Change view
Click to view available options
Quantity:
50 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC169580500 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC169580500 Supplier Thermo Scientific Chemicals Supplier No. 169580500

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 17814-85-6
Molecular Formula C23H24BrO2P
Molecular Weight (g/mol) 443.32
MDL Number MFCD00011906
InChI Key MLOSJPZSZWUDSK-UHFFFAOYSA-N
Synonym 4-carboxybutyl triphenylphosphonium bromide, phosphonium, 4-carboxybutyl triphenyl-, bromide, 4-carboxybutyl triphenylphosphanium bromide, carboxybutyltriphenylphosphonium bromide, 5-triphenylphosphonio pentanoic acid bromide, 4-carboxybutyl-triphenylphosphonium bromide, triphenyl 4-carboxybutyl phosphonium bromide, 4-carboxybutyl triphenyl phosphonium bromide
PubChem CID 161236
IUPAC Name (4-carboxybutyl)triphenylphosphanium bromide
SMILES [Br-].OC(=O)CCCC[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 204°C to 210°C
Color White
Loss on Drying 0.2% max.
Infrared Spectrum Authentic
Linear Formula HO2C(CH2)4P(C6H5)3Br
Quantity 50 g
Formula Weight 443.32
Percent Purity 97.5 to 102.5%
Physical Form Crystalline Powder
Chemical Name or Material (4-Carboxybutyl)triphenylphosphonium bromide, 98%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.