Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Carboxymethyl-6,7-methylenedioxycoumarin 95.0+%, TCI America™
SDP

Catalog No. C11961G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C11961G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C11961G Supplier TCI America Supplier No. C11961G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 97744-77-9
Molecular Formula C12H8O6
Molecular Weight (g/mol) 248.19
MDL Number MFCD00143278
InChI Key PIHRLFGNZWHKIO-UHFFFAOYSA-N
PubChem CID 44630205
IUPAC Name 2-{6-oxo-2H,6H-[1,3]dioxolo[4,5-g]chromen-8-yl}acetic acid
SMILES OC(=O)CC1=CC(=O)OC2=CC3=C(OCO3)C=C12

Specifications

Melting Point 195°C
Quantity 1 g
Formula Weight 248.19
Percent Purity ≥95.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Carboxymethyl-6,7-methylenedioxycoumarin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.