Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 4-Carboxyphenylboronic Acid

Catalog No. 45677210G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
45-677-210G 10 g
45-677-21G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 45-677-210G Supplier Sigma Aldrich Supplier No. 45677210G
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

4-Carboxyphenylboronic Acid

Specifications

CAS 14047-29-1
Melting Point 220°C
Molecular Formula C7H7BO4
Linear Formula HO2CC6H4B(OH)2
Quantity 10 g
MDL Number MFCD00151801
Synonym 4-(Dihydroxyboronyl)benzoic acid; 4-(Dihydroxyboryl)benzoic acid; 4-Boronobenzoic acid; 4-Carboxybenzeneboronic acid; 4-Carboxylphenylboronic acid; 4-Carboxylphenylboronic acid; 4-Hydroxycarbonylphenyl boronic acid; NSC 221170; p-Boronobenzoic acid; p-Carboxybenzeneboronic acid; p-Carboxyphenylboronic acid
Molecular Weight (g/mol) 165.94 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.