Learn More
Alkali Scientific Alkali Scientific™ 4" Cardboard Freezer Boxes with Drain Holes - Water and Ice Resistant Coating, w/ 36 Dividers, 50/Case
SDP
Catalog No. 503028726
Encompass_Preferred
• Choose optional slot/cell dividers• Dividers also sold separately, item CD-• Temperature Range: -196 to +121• Drain holes in bottom corners• Writing area on the lid• Keeps tubes in an upright position for easy sorting Alkali Scientific™ 4" cardboard freezer box are ideal forn any chemical laboratories, medicinal facilities or healthcare units for a wide range of purposes such as scientific research and development. Cardboard Freezer Box - 5 3/16x 5 3/16 x 4 15/16″ ( 148 x 148 x 125mm)
Catalog No. 50-302-8726 Supplier Alkali Scientific Supplier No. FB4C36C50
• Choose optional slot/cell dividers• Dividers also sold separately, item CD-• Temperature Range: -196 to +121• Drain holes in bottom corners• Writing area on the lid• Keeps tubes in an upright position for easy sorting Alkali Scientific™ 4" cardboard freezer box are ideal forn any chemical laboratories, medicinal facilities or healthcare units for a wide range of purposes such as scientific research and development. Cardboard Freezer Box - 5 3/16x 5 3/16 x 4 15/16″ ( 148 x 148 x 125mm)