Learn More
AdipoGen 4-CF3 umbelliferyl phosphate
SDP
Catalog No. 502560052
Encompass_Preferred
Chemical. CAS 352525-17-8. Formula C10H4F3Na2O6P. MW 354.08. Synthetic. Suitable as fluorogenic substrate for alkaline phosphatase.
Catalog No. 50-256-0052 Supplier AdipoGen Supplier No. CDXT0007M100
Chemical. CAS 352525-17-8. Formula C10H4F3Na2O6P. MW 354.08. Synthetic. Suitable as fluorogenic substrate for alkaline phosphatase.