Learn More
Cellvis 4-CH 35MM GL BTM 20 MW 1.5CG
Catalog No. NC0600518
Encompass
35 mm glass bottom dish with 4 chambers, 20mm microwell, #1.5 cover glass (0.16-0.19mm)
Catalog No. NC0600518 Supplier Cellvis Supplier No. D35C4201.5N
35 mm glass bottom dish with 4 chambers, 20mm microwell, #1.5 cover glass (0.16-0.19mm)