Learn More
Cellvis 4-CHAMBER 35MM GB DISH 20MM W/
Catalog No. NC0832919
Encompass
35 mm glass bottom dish with 4 chambers, 20mm microwell, #0 cover glass (0.085-0.115mm) A 4 Chamber 35mm glass bottom dish has the same size as a 35mm glass bottom dish. However the dish is divided into 4 chambers that quadruples sample size. •Suitable for long term tissue culture •Manufactured in a class 10,000 clean room •Dish made from virgin polystyrene, tissue culture treated. •German cover glass of superior optical quality • A USP class VI adhesive is used to assemble the cover glass and the dish. •Packed in easy to open peelable bag •Sterilized by Gamma radiation. 100/case
Catalog No. NC0832919 Supplier Cellvis Supplier No. D35C4200N
35 mm glass bottom dish with 4 chambers, 20mm microwell, #0 cover glass (0.085-0.115mm) A 4 Chamber 35mm glass bottom dish has the same size as a 35mm glass bottom dish. However the dish is divided into 4 chambers that quadruples sample size. •Suitable for long term tissue culture •Manufactured in a class 10,000 clean room •Dish made from virgin polystyrene, tissue culture treated. •German cover glass of superior optical quality • A USP class VI adhesive is used to assemble the cover glass and the dish. •Packed in easy to open peelable bag •Sterilized by Gamma radiation. 100/case