Learn More
Omega Engineering, Inc. Four-Channel Data Logger with USB and RS232
Catalog No. NC9003729
Encompass
The HH309A is a small 4-input thermometer plus a powerful data logger. The unit can store up to 16,000 records per channel at programmed intervals. The REC button is used to start and stop recording. You can store as many data sets as you want until memory is full. The unit comes with Windows-based software (Windows 98, NT, XP, ME, Windows 7) for the display and saving of data.
Catalog No. NC9003729 Supplier Omega Engineering, Inc. Supplier No. HH309A
The HH309A is a small 4-input thermometer plus a powerful data logger. The unit can store up to 16,000 records per channel at programmed intervals. The REC button is used to start and stop recording. You can store as many data sets as you want until memory is full. The unit comes with Windows-based software (Windows 98, NT, XP, ME, Windows 7) for the display and saving of data.