Learn More
World Precision Instrument 4-Channel Transducer Amplifier
SDP
Catalog No. 502538490
Encompass
Amplify output voltage signals
Transbridge (TBM4M) is a four-channel analog transducer manifold, specifically designed to amplify output voltage signals from pressure, force, displacement, and temperature transducers as well as a wide variety of other signal sources. Analog output signals are available from each channel for input to a data acquisition system for digital signal processing in a computer. Each channel contains a regulated 10 V power supply (+5 and -5 V with respect to signal ground) to provide DC power to transducers, and a precision differential amplifier with selectable voltage amplification and variable position adjustment control.
Catalog No. 50-253-8490 Supplier World Precision Instrument Supplier No. SYSTBM4M
Amplify output voltage signals
Transbridge (TBM4M) is a four-channel analog transducer manifold, specifically designed to amplify output voltage signals from pressure, force, displacement, and temperature transducers as well as a wide variety of other signal sources. Analog output signals are available from each channel for input to a data acquisition system for digital signal processing in a computer. Each channel contains a regulated 10 V power supply (+5 and -5 V with respect to signal ground) to provide DC power to transducers, and a precision differential amplifier with selectable voltage amplification and variable position adjustment control.