Learn More
Bulldog Bio Inc 4-CHIP DISP HEMOCYTOMETER NI
SDP
Catalog No. NC1211379
Encompass_Preferred
4-Chip Disposable Hemocytometer, NI, 50 Slides, 200 Tests. The 4-Chip is a new-style disposable plastic hemocytometer, that improves upon our popular C-Chip design by now providing twice as many counting chambers per chip. This doubling of tests is provided at almost the same price as the 2-chamber version, effectively cutting your lab’s cost by nearly half. The 4-Chip offers the ability to eliminate tedious washing and reduce exposure to infectious samples and contamination. 25 x 75 x 2 mm
Catalog No. NC1211379 Supplier Bulldog Bio Inc Supplier No. DHCN420
4-Chip Disposable Hemocytometer, NI, 50 Slides, 200 Tests. The 4-Chip is a new-style disposable plastic hemocytometer, that improves upon our popular C-Chip design by now providing twice as many counting chambers per chip. This doubling of tests is provided at almost the same price as the 2-chamber version, effectively cutting your lab’s cost by nearly half. The 4-Chip offers the ability to eliminate tedious washing and reduce exposure to infectious samples and contamination. 25 x 75 x 2 mm