Learn More
Bulldog Bio Inc 4-Chip Disposable Hemocytometer, NI, 10 Slides, 40 Tests
SDP
Catalog No. NC1731934
Encompass_Preferred
The 4-Chip is a new style of disposable plastic hemocytometer that improves upon our popular C-Chip design. With twice as many tests at almost the same price as the 2-chamber version, this product effectively cuts your lab’s cost by nearly half. It’s easier to use too! With a conventional glass hemocytometer, improper fit between coverslip and slide can change the volume of the sample in the chamber causing variability in counts.
Catalog No. NC1731934 Supplier Bulldog Bio Inc Supplier No. DHCN404
The 4-Chip is a new style of disposable plastic hemocytometer that improves upon our popular C-Chip design. With twice as many tests at almost the same price as the 2-chamber version, this product effectively cuts your lab’s cost by nearly half. It’s easier to use too! With a conventional glass hemocytometer, improper fit between coverslip and slide can change the volume of the sample in the chamber causing variability in counts.